Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylnaphthalene 95.0+%, TCI America™
SDP

Catalog No. I0327500ML
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 2027-17-0
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00014322
InChI Key TVYVQNHYIHAJTD-UHFFFAOYSA-N
Synonym 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene
PubChem CID 16238
IUPAC Name 2-propan-2-ylnaphthalene
SMILES CC(C)C1=CC2=CC=CC=C2C=C1

Specifications

Color Yellow
Boiling Point 268°C
Quantity 500 mL
Formula Weight 170.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropylnaphthalene
EINECSNumber (4)-0314&(4)-0961
RTECSNumber QJ8000000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.