Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanoethyl diisopropylchlorophosphoramidite, 95%, Thermo Scientific Chemicals

Catalog No. AAL1857506
Click to view available options
Quantity:
1 g
5 g

2-Cyanoethyl diisopropylchlorophosphoramidite is a useful intermediate in organic synthesis. It acts as a protecting reagent in DNA synthesis. It is involved in the selective monophosphorylation of carbohydrates and nucleosides. It plays an essential role in the conversion of protected ribonucleosides to phosphoramidites. It is also utilized as phosphitylating reagent for 3'-hydroxyl groups in the synthesis of oligodeoxyribonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanoethyl diisopropylchlorophosphoramidite is a useful intermediate in organic synthesis. It acts as a protecting reagent in DNA synthesis. It is involved in the selective monophosphorylation of carbohydrates and nucleosides. It plays an essential role in the conversion of protected ribonucleosides to phosphoramidites. It is also utilized as phosphitylating reagent for 3′-hydroxyl groups in the synthesis of oligodeoxyribonucleotides.

Solubility
Miscible with chloroform and dichloromethane.

Notes
Reacts violently with water. Incompatible with alcohols and strong bases.

Chemical Identifiers

CAS 89992-70-1
Molecular Formula C9H18ClN2OP
Molecular Weight (g/mol) 236.68
MDL Number MFCD00011544
InChI Key QWTBDIBOOIAZEF-UHFFFAOYSA-N
Synonym 2-cyanoethyl n,n-diisopropylchlorophosphoramidite, 2-cyanoethyl-n,n-diisopropylchlorophosphoramidite, phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethoxy diisopropylamino chlorophosphine, 3-chloro diisopropylamino phosphino oxy propanenitrile, 3-chloro-diisopropylamino phosphanyl oxypropanenitrile, 3-chloro diisopropylamino phosphanyl oxy propanenitrile, acmc-20anxd, 2-cyanoethyl diisopropylchloro-phosphoramidite, 2-cyanoethyl diisopropylphosphoramidochloridite
PubChem CID 2734844
IUPAC Name 3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile
SMILES CC(C)N(C(C)C)P(OCCC#N)Cl

Specifications

Density 1.061
Boiling Point 103°C to 105°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.478
Quantity 5 g
UN Number UN2845
Beilstein 3588525
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform and dichloromethane.
Formula Weight 236.68
Percent Purity 95%
Chemical Name or Material 2-Cyanoethyl diisopropylchlorophosphoramidite
Show More Show Less
Hazard Category H250-H314
Hazard Statement GHS H Statement
H250-H252-H314-H318-H302
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P222-P260-P264b-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.