Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-chloropurine riboside, 95%, Thermo Scientific Chemicals

Catalog No. AAJ6446803
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2004-07-1
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005735
InChI Key TXWHPSZYRUHEGT-UHFFFAOYNA-N
Synonym 2-amino-6-chloropurine riboside, 2-amino-6-chloropurine-9-riboside
PubChem CID 46874491
IUPAC Name (3R,4S,5R)-2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2Cl)N

Specifications

Melting Point 165°C to 167°C
Color White
Quantity 1 g
Formula Weight 301.69
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Amino-6-chloropurine riboside
EINECSNumber 217-905-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.