Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. N09975G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 15513-52-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00023503
InChI Key AETHUDGJSSKZKT-UHFFFAOYSA-N
Synonym 3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
PubChem CID 84958
IUPAC Name 2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])C

Specifications

Melting Point 38°C
Color Yellow
Boiling Point 105°C
Quantity 5 g
UN Number 1325
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-3-nitropyridine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.