Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1268236
Click to view available options
Quantity:
500 g

2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

Solubility
Soluble in water <0.1 g/100 mL at 18°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 95-78-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007743
InChI Key VOWZNBNDMFLQGM-UHFFFAOYSA-N
Synonym 2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
PubChem CID 7259
ChEBI CHEBI:518305
IUPAC Name 2,5-dimethylaniline
SMILES CC1=CC=C(C)C(N)=C1

Specifications

Melting Point 15°C
Density 0.973
Boiling Point 218°C
Quantity 500 g
Odor Characteristic
Refractive Index 1.559
UN Number UN1711
Beilstein 2205178
Merck Index 14,10084
Solubility Information Soluble in water <0.1g/100 mL at 18°C.
Formula Weight 121.18
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylaniline
Show More Show Less
Hazard Category H227-H301+H311-H330-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID
EINECSNumber 202-451-0
RTECSNumber ZE9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.