Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3(2H)-furanone, 94%, Thermo Scientific Chemicals

Catalog No. AAA1218214
Click to view available options
Quantity:
1g
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14400-67-0
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00052571
InChI Key ASOSVCXGWPDUGN-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3 2h-furanone, 2,5-dimethylfuran-3 2h-one, 2,5-dimethyl-2,3-dihydrofuran-3-one, 3 2h-furanone, 2,5-dimethyl, 2,5-dimethyl-3 2h furanone, 2,5-dimethyl-2h-furan-3-one, 2,5-dimethyl-3-2h-furanone, 2h-furan-3-one, 2,5-dimethyl, mango furanone
PubChem CID 85730
IUPAC Name 2,5-dimethylfuran-3-one
SMILES CC1C(=O)C=C(O1)C

Specifications

Density 1.06
Boiling Point 259°C to 261°C
Flash Point 68°C (154°F)
Odor Sweet
Quantity 25 g
Formula Weight 112.13
Percent Purity 94%
Chemical Name or Material 2,5-Dimethyl-3 (2H)-furanone
Hazard Category H227-H302
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 238-365-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.