Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine 95.0+%, TCI America™
SDP

Catalog No. B45871G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 851030-18-7
Molecular Formula C17H14ClN3O2
Molecular Weight (g/mol) 327.77
MDL Number MFCD29089341
InChI Key XKDFUXJROYAXAO-UHFFFAOYSA-N
Synonym DBT-Cl
PubChem CID 71406442
IUPAC Name 2,4-bis(benzyloxy)-6-chloro-1,3,5-triazine
SMILES ClC1=NC(OCC2=CC=CC=C2)=NC(OCC2=CC=CC=C2)=N1

Specifications

Melting Point 66°C
Color White
Quantity 1 g
Formula Weight 327.77
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.