Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride 97.0+%, TCI America™
SDP

Catalog No. B03251G
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 38800-20-3
Molecular Formula C17H13ClN4S
Molecular Weight (g/mol) 340.829
MDL Number MFCD00060088
InChI Key RLHVMOZOYHDIGV-UHFFFAOYSA-M
Synonym STC
PubChem CID 6097060
IUPAC Name 2,3-diphenyl-5-thiophen-2-yltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CS4.[Cl-]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 340.83
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.