Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromo-4-methoxyphenyl)ethylamine, 98%, Thermo Scientific™

Catalog No. AAL1987403
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 159465-27-7
Molecular Formula C9H12BrNO
Molecular Weight (g/mol) 230.105
MDL Number MFCD00153891
InChI Key JHVALSRTUOVNLL-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxyphenethylamine, 2-3-bromo-4-methoxyphenyl ethanamine, 2-3-bromo-4-methoxyphenyl ethylamine, acmc-20an8n, 3-bromo-4-methoxy-phenethyl amine, benzeneethanamine,3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethaneamine, 2-3-bromo-4-methoxyphenyl-ethylamine, 2-3-bromo-4-methoxy-phenyl-ethylamine
PubChem CID 2735581
IUPAC Name 2-(3-bromo-4-methoxyphenyl)ethanamine
SMILES COC1=C(C=C(C=C1)CCN)Br

Specifications

Density 1.389
Boiling Point 114°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.58
Quantity 1 g
UN Number UN2735
Beilstein 3250744
Formula Weight 230.11
Percent Purity 98%
Chemical Name or Material 2-(3-Bromo-4-methoxyphenyl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.