Learn More
Alkali Scientific CellPro™ 1X DPBS, Dulbecco’s Phosphate Buffered Saline, without Calcium or Magnesium, Sterile Filtered
Supplier: Alkali Scientific PBD1000
Like PBS, DPBS is also a water-based solution containing sodium chloride and phosphate buffer that is used in biological applications. However, DPBS differs from PBS in that it also includes potassium chloride and is available in a larger variety of formulations.