Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%, Thermo Scientific Chemicals

Catalog No. AAL1659914
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 203303-01-9
Molecular Formula C9H6F14O2
Molecular Weight (g/mol) 412.123
MDL Number MFCD00153275
InChI Key XQULMKMNFZLURS-UHFFFAOYSA-N
Synonym 1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
PubChem CID 2776243
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
SMILES C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 108°C to 112°C
Boiling Point 140°C (5 mmHg)
Linear Formula HOCH2(CF2)7CH2OH
Quantity 25 g
Formula Weight 412.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 1H,1H,9H,9H-Perfluoro-1,9-nonanediol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.