Learn More
Glass Expansion 19 mm Metal Ball Joint Clip
Supplier: Glass Expansion JC19
19mm Metal Ball Joint Clip. Used to secure 19mm male and female ball joints for torches, transfer tubes and spray chambers. For 13mm ball joints use JC-13.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001