Learn More
Sarstedt Inc 16.5X95 TUBE 12MLFB GWB/PK1000
Supplier:  Sarstedt Inc 62.470.300
Encompass_Preferred
12ml Screen Printed Tube 16.5x95mm Flat Bottom Polystyrene Graduations and Writing Block No Cap
Catalog No. NC9170747
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.