Learn More
Andwin Scientific 15ML DOUNCE HOMOGENIZER
SDP
Supplier:  Andwin Scientific AX24835B
Encompass_Preferred
15 ML TISSUE GRINDER 2/CS, Dounce Grinders are designed to avoid damage to nuclei and mitochondria while reducing soft tissue. Ideal for enzyme studies. Includes mortar with reservoir and pourout, and both loose and tight pestles,Borosilica
Catalog No. NC9661231
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.