Learn More
Chemglass Life Sciences 13-425 G.P.I. Glass Thread, 12mm OD Tubing, 100mm OAL
SDP
Supplier:  Chemglass Life Sciences CG19002
Encompass_Preferred
13-425 G.P.I. Glass Thread, 12mm OD Tubing, 100mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.F129:G137
Catalog No. CG19002
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.