Learn More
Andwin Scientific 12 X 75 MM PP CULTURE TUBE BU
Supplier: Andwin Scientific 14500000
The 12 x 75 mm tubes are made with polypropylene. The tubes do not come with caps but are compatible with polyethylene plug caps (1494-0000) or dual position caps (1494-1030), which are both sold separately. It is sold in a pack of 1000 tubes.