Learn More
Quality Biological Inc TBE Buffer 10X, CS 10 X 1000 mL
SDP
Supplier:  Quality Biological Inc 351001131CS
Encompass_Preferred
TBE (TRIS Borate EDTA) is often used in procedures involving nucleic acids, the most common being electrophoresis. • Tris-acid solutions are effective buffers for slightly basic conditions, which keep DNA deprotonated and soluble in water. • Applications: TBE Buffer, 1X is used for DNA electrophoresis. • TBE Buffer, 0.5X is used for electrophoresis onto DEAE cellulose membrane. • TBE Buffer, 2X is used for polyacrylamide and agarose electrophoresis. • TBE is also used for other nucleic acid electrophoresis procedures such as Northern and Southern blots. • Storage Conditions: 15 to 30°C • Shipping Condition: ambient temperature • Sterile Filtered • DNase, RNase, and Protease Tested • Product and/or packaging contains recycled/renewable content • Product and/or packaging is recyclable or reduces waste • Product and/or packaging is safer to human and environmental health • Harmonization Code: 3822.00.5090 • ECCN: EAR99 • Max Shelf Life: 36 months
Catalog No. 50-751-7294
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.