Learn More
Chemglass Life Sciences 100mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 41 O.D. x 225mm Height
SDP
Supplier:  Chemglass Life Sciences AF008405
Encompass_Preferred
100mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 41 O.D. x 225mm Height. Round bottom, cylindrical drying ampuls with 10mm OD top stem. Manufactured from Type 1 borosilicate glass. Please note: Ampuls are available with top stems to fit AF-0067 Cajon Unions.
Catalog No. 50-872-929
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.