Learn More
New England Biolabs, Inc. NEB 10-beta Competent E. coli (High Efficiency) – 1 96-well plate, 20uL/well
Supplier: New England Biolabs, Inc. C3019P
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high- quality plasmid preparations.