Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-4-(trifluoromethylthio)benzene, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1248503
Click to view available options
Quantity:
1 g

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 940-76-1
Molecular Formula C7H4F4S
Molecular Weight (g/mol) 196.163
MDL Number MFCD00040978
InChI Key BTKQCAUEXFTVNT-UHFFFAOYSA-N
Synonym 1-fluoro-4-trifluoromethylthio benzene, 4-fluorophenyl trifluoromethyl sulphide, 1-fluoro-4-trifluoromethylsulfanyl benzene, 1-fluoro-4-trifluoromethyl sulfanyl benzene, trifluoro 4-fluorophenylthio methane, p-fluorophenyltrifluoromethyl sulfide, 1-trifluoromethylthio-4-fluorobenzene, 1-fluoro-4-trifluoromethyl thio benzene, 1-fluoro-4-trifluoromethyl thio-benzene
PubChem CID 136758
IUPAC Name 1-fluoro-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1F)SC(F)(F)F

Specifications

Density 1.38
Boiling Point 136°C to 138°C
Flash Point 37°C (99°F)
Odor Unpleasant
Refractive Index 1.45
Quantity 1 g
UN Number UN1993
Beilstein 2046333
Formula Weight 196.17
Percent Purity ≥98%
Chemical Name or Material 1-Fluoro-4-(trifluoromethylthio)benzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.