Learn More
Andwin Scientific 1.5 ML SEAL-RITE MICROCENTRIF
Supplier: Andwin Scientific 16155501
The Seal-Rite 1.5 mL microcentrifuge tube protects samples from evaporation and spillage with its dependable precision sealing system. The tubes will remain tightly closed during the processes of boiling, freezing, centrifugation, and exposure to phenol or chloroform mixture, but can be opened with no difficulty by hand. This product is free of RNase, DNase, DNA, and pyrogens.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.