Learn More
Andwin Scientific 1.5 ML MICROCENTRIFUGE TUBE P
Supplier: Andwin Scientific 14151448
The 1.5 ml microcentrifuge tube has an attached flat cap that makes this product ideal for boiling. It also has graduations every 100 l that allow precise sample measurement. The tube is made with polypropylene and is free of RNase, DNase, DNA, and pyrogen. It comes in a bag of 500 tubes in assorted neon colors of blue, green, orange, red, violet, and yellow.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.