Learn More
Chemglass Life Sciences Adapter, Flexible, Cajon Ultra-Torr, 24in Flexible Length, Airfree, Schlenk, For Use with 1/4in OD Tube
Supplier: Chemglass Life Sciences AF006611
Adapter, Flexible, Cajon Ultra-Torr, 24in Flexible Length, Airfree, Schlenk, For Use with 1/4in OD Tube. Cajon adapters provide the flexibility of rubber tubing and are the ideal replacement for rubber, plastic, or glass tubing in critical vacuum applications. Connections compensate for misalignment, expansion and contraction; they are compressible by at least 20%, and they are extendable by 50% of its nominal listed length. Cajon adapters are supplied complete with two AF-0067 unions, two AF-0068 cuff adapters, and listed length of flexible tubing. NOTE: Since Flexible tubing will contract when evacuated, glass apparatus should NOT be firmly clamped in place until vacuum is established within the system.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.