Learn More
Andwin Scientific 1.3CM TESTTUBE BRUSH NAT 10/PK
SDP
Supplier:  Andwin Scientific 17027023
Encompass_Preferred
The Test Tube Brushes feature a radial tip that can efficiently clean test tubes and narrow mouth glassware. Its tip also offers protection of glasswares. The brush has an overall length of 20 centimeters and its brush head has a dimension of 3 inches. The brush's bristles are made of nylon. It is sold in a pack of 10.
Catalog No. NC9600595
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.