Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-2,2-dimethoxypropane, 98%, Thermo Scientific Chemicals

Catalog No. AAA1121714
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22094-18-4
Molecular Formula C5H10Br2O2
Molecular Weight (g/mol) 261.941
MDL Number MFCD00051792
InChI Key CPAHOXOBYHMHDT-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320
PubChem CID 2734196
IUPAC Name 1,3-dibromo-2,2-dimethoxypropane
SMILES COC(CBr)(CBr)OC

Specifications

Melting Point 64°C to 67°C
Quantity 25 g
Beilstein 1698992
Formula Weight 261.95
Percent Purity 99%
Chemical Name or Material 1,3-Dibromo-2,2-dimethoxypropane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.