Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%, Thermo Scientific™

Catalog No. AAL0443618
Click to view available options
Quantity:
50 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-Bis(chloromethyl)tetramethyldisiloxane is a useful bifunctional silylating agent.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2362-10-9
MDL Number MFCD00000877

Specifications

Boiling Point 92°C to 93°C (20mmHg)
Quantity 50 g
Molecular Formula C7H7BrO
Beilstein 506631
Flash Point 73°C (163°F)
Synonym 4-bromo-o-cresol, 2-methyl-4-bromophenol, 4-bromo-2-methyl-phenol, o-cresol, 4-bromo, phenol, 4-bromo-2-methyl, 2-hydroxy-5-bromotoluene, p-bromocresol, 4-bromo-cresol, pubchem7106, acmc-1chy8
Solubility Information Not miscible or difficult to mix with water.
InChI Key IWJGMJHAIUBWKT-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Br)O
IUPAC Name 4-bromo-2-methylphenol
Molecular Weight (g/mol) 187.04
PubChem CID 16896
Formula Weight 231.27
Percent Purity 97%
Refractive Index 1.44
Density 1.05
Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 219-109-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.