Learn More
Andwin Scientific 1.2ML INDIVIDUAL TUBES 1000/PK
SDP
Supplier:  Andwin Scientific P87011
Encompass_Preferred
The Micro Dilution tubes have a volume capacity of 1.2 mL. These tubes are free of RNase, DNase, and pyrogen. This is sold in a pack of 1000. The racks and the caps for these tubes are sold separately.
Catalog No. 50-999-411
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.