Learn More
MTC Bio ClearSeal™ screw-cap microtubes, 0.5mL, w/ O-ring & caps, sterile, non-graduated, self-standing, 20 resealable bags of 50 tubes, 1000/cs
Supplier:  MTC Bio C3205S
Encompass_Preferred
ClearSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0mL. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg*.
Catalog No. NC2017384
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.