Learn More
Chemglass Life Sciences 0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum, Perfluoro O-Ring
SDP
Supplier:  Chemglass Life Sciences CG96150
Encompass_Preferred
0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum, Perfluoro O-Ring. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.
Catalog No. 50-153-1870
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.