Learn More
Andwin Scientific 0.2ML 8-STRIP PCR TUBES 125/PK
Supplier: Andwin Scientific T30142
The 8-strip PCR tubes features a thin-wall with dimensional uniformity throughout the sidewall and the conical bottom. This product has detached caps. The tubes have a volume capacity of 0.2 mL. This is sold in a pack of 125.