Learn More
Andwin Scientific 0.2 ML INDIVIDUAL PCR TUBES WI
SDP
Supplier:  Andwin Scientific 14028108
Encompass_Preferred
Made with polypropylene thin walls, the 02. mL TempAssure PCR tubes are ideal for laboratory procedures that require quick and uniform heat transfer. This product is free of DNase, RNAse, DNA, PCR, and pyrogen. The tubes can be sealed with the attached flat cap that can be written on for easy sample identification. It comes in a bag of 1000 tubes in various colors of blue, green, red, violet, and yellow.
Catalog No. 50-819-652
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.