Learn More
Andwin Scientific 0.2 ML INDIVIDUAL PCR TUBES WI
Supplier: Andwin Scientific 14024300
Made with polypropylene conical walls that are up to 20% thinner than other brands, the 02. mL TempAssure PCR tubes are ideal for laboratory procedures that require quick and uniform heat transfer. This product is free of DNase, RNAse, DNA, PCR, and pyrogen. Each tube has an attached dome cap. It comes in a bag of 1000.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.