Learn More
Andwin Scientific 0.2 ML 8-TUBE STRIPS WITHOUT C
Supplier: Andwin Scientific 14023500
The 0.2 ml PCR 8-tube strip are specially designed with polypropylene thin wall to allow quick and even heat transfer for reliable amplifications. This product is free of detectable levels of DNase, RNase, DNA, pyrogen, and PCR inhibitors. It is sold in a pack of 125.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.