Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Barrel Only, Without Sidearm, 0-12mm Bore, 22mm OD Barrel
Supplier: Chemglass Life Sciences CG99803
Valve, Chem-Vac, Glass Barrel Only, Without Sidearm, 0-12mm Bore, 22mm OD Barrel. Glass barrel only, without sidearm, for all CHEM-VAC valves. Heavy duty rounded coarse thread and precision formed tapered seat. PLEASE NOTE: THE 0-20mm BORE SIZE VALVE IS Chemglass's PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.