Learn More
Hach Company Conductivity Standard Solution, 1412 µS/cm, KCl, 500 mL
Supplier: Hach Company 2974349
Conductivity Standard Solution. 1412 µS/cm ±10% at 25°C, 745.5 mg/L ±10% as Potassium Chloride (KCl) (0.01M). 500 mL bottle.